Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1foo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138311.2 | NP_612184.1 | 1, 2 | 148 | ||
GenBank mRNA | AK135944.1 | - | 1, 2 | 152 | ||
AK162137.1 | - | 1, 2 | 127 | |||
AK162154.1 | - | 1, 2 | 145 | |||
AY007195.1 | - | 1, 2 | 148 | |||
BC137916.1 | - | 1, 2 | 144 | 8860935 | ||
BC145046.1 | - | 1, 2 | 144 |