Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1foo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138311.2 | NP_612184.1 | 5 | 900 | ||
GenBank mRNA | AK135944.1 | - | 5 | 904 | ||
AK162137.1 | - | 5 | 879 | |||
AK162154.1 | - | 5 | 901 | |||
AK165127.1 | - | 6 | 982 | |||
AY007195.1 | - | 5 | 900 | |||
BC137916.1 | - | 5 | 896 | 8860935 | ||
BC145046.1 | - | 5 | 900 |