Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133930.1 | NP_598691.1 | 10 | 1488 | ||
GenBank mRNA | AB009020.1 | - | 10 | 1493 | ||
AK050160.1 | - | 3 | 269 | |||
AK050455.1 | - | 3 | 269 | |||
AK077726.1 | - | 10 | 1633 | |||
AK155777.1 | - | 10 | 1498 | |||
AK186101.1 | - | 1 | 88 | |||
AK208796.1 | - | 1 | 72 | |||
BC010804.1 | - | 10 | 1488 | 4239786 | ||
BC023058.1 | - | 2 | 794 | |||
BC023893.1 | - | 10 | 1499 | 5310111 | ||
BC025932.1 | - | 10 | 1394 | 5102571 | ||
BC029065.1 | - | 10 | 1524 | 4935296 |