Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138313.3 | NP_612186.2 | 4 | 681 | ||
GenBank mRNA | AK040622.1 | - | 4 | 638 | ||
AK084658.1 | - | 4 | 691 | |||
AK089218.1 | - | 3 | 604 | |||
AK169903.1 | - | 4 | 639 | |||
AK171688.1 | - | 1 | 40 | |||
AY029253.1 | - | 2 | 407 | |||
BC079650.1 | - | 4 | 695 | 6847314 |