Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285853.1 | NP_001272782.1 | 5 | 600 | ||
NM_001285854.1 | NP_001272783.1 | 5 | 597 | |||
NM_001285855.1 | NP_001272784.1 | 5 | 665 | |||
NM_010768.2 | NP_034898.1 | 6 | 733 | |||
GenBank mRNA | AK013606.1 | - | 5 | 414 | ||
AK018182.1 | - | 5 | 451 | |||
AK079816.1 | - | 5 | 393 | |||
AK158883.1 | - | 5 | 458 | |||
BC060086.1 | - | 5 | 554 | |||
BC111864.1 | - | 5 | 411 | 40046807 | ||
D45243.1 | - | 5 | 665 | |||
L27738.1 | - | 6 | 487 | |||
S77473.1 | - | 5 | 412 | |||
U05210.1 | - | 5 | 386 |