Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008568.2 | NP_032594.1 | 13 | 1892 | ||
GenBank mRNA | AK076059.1 | - | 9 | 1342 | ||
AK077766.1 | - | 10 | 1451 | |||
AK132748.1 | - | 2456 | ||||
AK133091.1 | - | 13 | 1888 | |||
AK144980.1 | - | 13 | 1904 | |||
AK149913.1 | - | 13 | 1886 | |||
AK150059.1 | - | 13 | 1889 | |||
AK150066.1 | - | 13 | 1886 | |||
AK154052.1 | - | 13 | 1889 | |||
BC023232.1 | - | 13 | 1868 | |||
BC065164.1 | - | 13 | 1876 | 6829737 | ||
BC066024.1 | - | 13 | 1831 | 6829337 | ||
D26091.1 | - | 13 | 1892 |