Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 18 | 2068 | ||
GenBank mRNA | AK010498.1 | - | 18 | 2035 | ||
AK011932.1 | - | 18 | 2068 | |||
AK129076.1 | - | 18 | 2021 | |||
AK137000.1 | - | 18 | 2095 | |||
AK145021.1 | - | 18 | 2003 | |||
AK145316.1 | - | 18 | 1977 | |||
AK161029.1 | - | 18 | 2077 | |||
AK164138.1 | - | 18 | 2035 | |||
AK167457.1 | - | 18 | 1992 | |||
BC020416.1 | - | 17 | 1903 | |||
BC085276.1 | - | 18 | 2101 | 30443236 | ||
L76158.1 | - | 18 | 1999 | |||
X95351.1 | - | 18 | 2006 |