Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Men1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168488.1 | NP_001161960.1 | 10 | 1593 | ||
NM_001168489.1 | NP_001161961.1 | 10 | 1571 | |||
NM_001168490.1 | NP_001161962.1 | 8 | 1742 | |||
NM_008583.2 | NP_032609.1 | 9 | 1907 | |||
GenBank mRNA | AB023401.1 | - | 9 | 1534 | ||
AF016398.1 | - | 10 | 1546 | |||
AF072755.1 | - | 10 | 1530 | |||
AF109389.1 | - | 9 | 1906 | |||
AF130368.1 | - | 9 | 1889 | |||
AK133984.1 | - | 10 | 1530 | |||
AK154371.1 | - | 10 | 1552 | |||
AK164700.1 | - | 10 | 1527 | |||
AK170713.1 | - | 9 | 1887 | |||
BC006610.1 | - | 9 | 1932 | |||
BC036287.1 | - | 8 | 1670 | 4189611 |