Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045489.1 | NP_001038954.1 | 6 | 783 | ||
NM_008594.2 | NP_032620.2 | 7 | 894 | |||
GenBank mRNA | AB021130.2 | - | 7 | 801 | ||
AB025280.1 | - | 6 | 690 | |||
AK087555.1 | - | 6 | 773 | |||
AK089211.1 | - | 6 | 749 | |||
AK145183.1 | - | 6 | 747 | |||
AK148371.1 | - | 6 | 772 | |||
AK152088.1 | - | 7 | 883 | |||
AK155047.1 | - | 7 | 892 | |||
AK169997.1 | - | 6 | 747 | |||
AK170935.1 | - | 6 | 747 | |||
AK171143.1 | - | 7 | 858 | |||
AK189618.1 | - | 2 | 168 | |||
AK207561.1 | - | 1 | 40 | |||
AK212128.1 | - | 1 | 40 | |||
AK212659.1 | - | 1 | 40 | |||
AK213407.1 | - | 1 | 40 | |||
AK214404.1 | - | 1 | 40 | |||
AK219710.1 | - | 1 | 40 | |||
BC003892.1 | - | 6 | 727 | 3593310 | ||
BC003904.1 | - | 6 | 722 | 3601565 | ||
BC018577.1 | - | 6 | 720 | |||
BC037475.1 | - | 3 | 1109 | |||
M38337.1 | - | 7 | 846 | |||
Y11684.2 | - | 6 | 690 |