Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008595.2 | NP_032621.1 | 5, 6 | 831 | ||
GenBank mRNA | AF015769.1 | - | 5, 6 | 658 | ||
AK143523.1 | - | 5, 6 | 865 | |||
BC010983.1 | - | 5, 6 | 853 | 3980692 | ||
U94349.1 | - | 5, 6 | 831 |