Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039104.2 | NP_001034193.2 | 13 | 1856 | ||
NM_018752.3 | NP_061222.3 | 13 | 1749 | |||
GenBank mRNA | AF047714.1 | - | 13 | 1618 | ||
AK044507.1 | - | 13 | 1780 | |||
AK148193.1 | - | 11 | 1698 | |||
AK148268.1 | - | 13 | 1603 | |||
AK148275.1 | - | 13 | 1659 | |||
AK149290.1 | - | 13 | 1778 | |||
AK189982.1 | - | 3 | 177 | |||
AK190619.1 | - | 3 | 286 | |||
AK192027.1 | - | 3 | 177 | |||
AK194126.1 | - | 3 | 177 | |||
AK196486.1 | - | 3 | 177 | |||
AK199395.1 | - | 3 | 177 | |||
AK199874.1 | - | 3 | 177 | |||
AK202089.1 | - | 3 | 182 | |||
AK202317.1 | - | 3 | 177 | |||
AK203960.1 | - | 3 | 177 | |||
AK205137.1 | - | 3 | 183 | |||
AK217903.1 | - | 3 | 178 | |||
AK219280.1 | - | 3 | 178 | |||
AY180104.2 | - | 13 | 1856 |