Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010818.3 | NP_034948.3 | 3, 4 | 597 | ||
GenBank mRNA | AF231126.1 | - | 2, 3 | 403 | ||
AK148308.1 | - | 3, 4 | 566 | |||
AK164478.1 | - | 3, 4 | 520 | |||
BC051984.1 | - | 3, 4 | 515 | 5707683 | ||
BC054759.1 | - | 3, 4 | 530 | 6413363 |