Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010835.2 | NP_034965.2 | 2 | 1123 | ||
GenBank mRNA | AF308572.1 | - | 2 | 907 | ||
AK077524.1 | - | 2 | 1126 | |||
AK078600.1 | - | 2 | 1124 | |||
AK145726.1 | - | 2 | 1130 | |||
BC016426.1 | - | 2 | 1106 | 4923403 | ||
X14759.1 | - | 2 | 992 | |||
X59251.1 | - | 2 | 982 |