Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010835.2 | NP_034965.2 | 1 | 420 | ||
GenBank mRNA | AF308572.1 | - | 1 | 204 | ||
AK077524.1 | - | 1 | 419 | |||
AK078600.1 | - | 1 | 420 | |||
AK081534.1 | - | 1 | 419 | |||
AK145726.1 | - | 1 | 418 | |||
BC016426.1 | - | 1 | 403 | 4923403 | ||
X14759.1 | - | 1 | 291 | |||
X59251.1 | - | 1 | 291 |