Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021296.2 | NP_067271.1 | 2 | 130 | ||
GenBank mRNA | AF041060.1 | - | 2 | 95 | ||
AK003011.1 | - | 2 | 111 | |||
AK012634.1 | - | 2 | 109 | |||
AK082983.1 | - | 120 | ||||
AK134455.1 | - | 2 | 109 | |||
AK145493.1 | - | 2 | 122 | |||
BC068232.1 | - | 2 | 107 | 30533079 | ||
BC094583.1 | - | 2 | 97 | |||
BC120747.1 | - | 2 | 120 | 40129670 | ||
BY249944.1 | - | 2 | 156 |