Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039934.1 | NP_001035023.1 | 6 | 635 | ||
NM_008632.2 | NP_032658.2 | 6 | 635 | |||
GenBank mRNA | AK046341.1 | - | 4 | 500 | ||
AK079618.1 | - | 4 | 664 | |||
AK086484.1 | - | 4 | 499 | |||
AK141458.1 | - | 4 | 400 | |||
AK166380.1 | - | 6 | 651 | |||
BC048781.1 | - | 2 | 333 | |||
BC051410.1 | - | 2 | 330 | |||
BC052446.1 | - | 6 | 635 | |||
BC150945.1 | - | 6 | 634 | 9087856 | ||
BU056737.1 | - | 6 | 635 | |||
CN538992.1 | - | 2 | 280 | |||
M21041.1 | - | 4 | 401 |