Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043355.2 | NP_001036820.2 | 1 | 357 | ||
NM_001048167.1 | NP_001041632.1 | 1 | 1256 | |||
NM_010837.3 | NP_034967.2 | 1 | 1256 | |||
GenBank mRNA | AK018193.2 | - | 1 | 736 | ||
AK082709.1 | - | 1 | 1215 | |||
AK087413.1 | - | 1 | 1219 | |||
AK154455.1 | - | 1 | 329 | |||
BC053039.1 | - | 1 | 1257 | 5709455 | ||
Y16032.1 | - | 1 | 297 |