Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253877.1 | NP_001240806.1 | 9 | 1426 | ||
NM_001253878.1 | NP_001240807.1 | 9 | 1145 | |||
NM_001253879.1 | NP_001240808.1 | 8 | 1068 | |||
NM_001253880.1 | NP_001240809.1 | 6 | 783 | |||
NM_013827.3 | NP_038855.2 | 8 | 1064 | |||
GenBank mRNA | AB221622.1 | - | 5 | 471 | ||
AK030752.1 | - | 8 | 1014 | |||
AK031409.1 | - | 10 | 1133 | |||
AK032798.1 | - | 9 | 1387 | |||
AK032819.1 | - | 9 | 1388 | |||
AK137539.1 | - | 10 | 1373 | |||
AK139685.1 | - | 9 | 1146 | |||
BC024889.1 | - | 8 | 1036 | |||
BC029076.1 | - | 8 | 975 | |||
BC050826.1 | - | 8 | 1018 | |||
BC057163.1 | - | 8 | 861 | |||
BC071223.1 | - | 6 | 536 | |||
BC092237.1 | - | 8 | 996 | 30637248 | ||
BC100340.1 | - | 6 | 714 | 30715034 | ||
S78454.1 | - | 5 | 471 |