Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039657.2 | NP_001034746.1 | 1 | 222 | ||
NM_001039658.2 | NP_001034747.1 | 1 | 222 | |||
GenBank mRNA | AB057423.1 | - | 1 | 210 | ||
AK015724.1 | - | 1 | 163 | |||
AK015732.1 | - | 1 | 195 | |||
AK053809.1 | - | 1 | 195 | |||
AK161511.1 | - | 1 | 223 |