Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039657.2 | NP_001034746.1 | 1 | 403 | ||
NM_001039658.2 | NP_001034747.1 | 1 | 403 | |||
GenBank mRNA | AB057423.1 | - | 1 | 391 | ||
AK015724.1 | - | 1 | 344 | |||
AK015732.1 | - | 1 | 376 | |||
AK053809.1 | - | 1 | 376 | |||
AK161511.1 | - | 1 | 404 | |||
BC024377.1 | - | 1 | 379 | 4976492 | ||
U77383.1 | - | 1 | 40 |