Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023166.2 | NP_075655.2 | 1 | 347 | ||
NM_181452.2 | NP_852117.2 | 1 | 347 | |||
GenBank mRNA | AB012726.1 | - | 1 | 332 | ||
AF212920.1 | - | 1 | 329 | |||
AF212921.1 | - | 1 | 314 | |||
AK003883.1 | - | 1 | 335 | |||
AK013256.1 | - | 1 | 314 | |||
BC013894.1 | - | 1 | 325 | 4217979 | ||
BC030163.1 | - | 1 | 318 | 3674980 | ||
BC056166.1 | - | 1 | 332 | 5715229 | ||
BM942073.1 | - | 1 | 273 | |||
CJ132526.1 | - | 1 | 347 |