Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144514.2 | NP_653097.2 | 1 | 191 | ||
GenBank mRNA | AK082909.1 | - | 1 | 168 | ||
AK133507.1 | - | 1 | 167 | |||
AK147766.1 | - | 29 | ||||
BC030052.1 | - | 1 | 91 | |||
BC038641.1 | - | 1 | 138 | |||
BC051210.1 | - | 1 | 152 | |||
BC132083.1 | - | 1 | 180 | 40130360 | ||
BC132085.1 | - | 1 | 181 | 40130362 | ||
BY337798.1 | - | 1 | 192 | |||
D85430.1 | - | 1 | 161 |