Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013606.1 | NP_038634.1 | 13 | 1829 | ||
GenBank mRNA | AB029920.1 | - | 13 | 1829 | ||
AK143831.1 | - | 13 | 1833 | |||
AK147949.1 | - | 2452 | ||||
BC007127.1 | - | 13 | 1814 | 3591798 | ||
J03368.1 | - | 13 | 1830 |