Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008542.2 | NP_001008542.2 | 4, 5 | 740 | ||
NM_001008543.2 | NP_001008543.1 | 4, 5 | 427 | |||
NM_010847.3 | NP_034977.1 | 4, 5 | 580 | |||
GenBank mRNA | AK140465.1 | - | 4, 5 | 530 | ||
AK144219.1 | - | 4, 5 | 622 | |||
AK154538.1 | - | 4, 5 | 730 | |||
BC064453.1 | - | 4, 5 | 725 | 6810025 | ||
D31824.1 | - | 4, 5 | 580 | |||
L38821.1 | - | 4, 5 | 433 | |||
L38822.1 | - | 4, 5 | 452 | |||
U24673.1 | - | 4, 5 | 413 | |||
U24674.1 | - | 4, 5 | 362 |