Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010851.2 | NP_034981.1 | 5 | 896 | ||
GenBank mRNA | AK090107.1 | - | 3 | 1408 | ||
AK150083.1 | - | 5 | 779 | |||
AK151748.1 | - | 5 | 852 | |||
AK152596.1 | - | 5 | 881 | |||
AK152935.1 | - | 5 | 854 | |||
AK167929.1 | - | 5 | 876 | |||
BC005591.1 | - | 5 | 896 | 3482246 | ||
BC051075.1 | - | 5 | 843 | |||
BC058787.1 | - | 5 | 831 | 6309465 | ||
DQ790885.1 | - | 5 | 815 | |||
DQ790886.1 | - | 5 | 815 | |||
U84409.1 | - | 5 | 877 | |||
X51397.1 | - | 5 | 765 |