Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162417.1 | NP_001155889.1 | 13 | 1401 | ||
NM_001162418.1 | NP_001155890.1 | 12 | 1350 | |||
NM_010852.2 | NP_034982.2 | 12 | 1350 | |||
GenBank mRNA | AF483504.1 | - | 11 | 1076 | ||
AF483505.1 | - | 11 | 1076 | |||
AK012133.1 | - | 12 | 1352 | |||
AK048404.1 | - | 13 | 1368 | |||
AK129336.1 | - | 13 | 1313 | |||
AK143230.1 | - | 12 | 1286 | |||
U13262.1 | - | 8 | 791 |