Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022879.2 | NP_075017.2 | 5 | 337 | ||
GenBank mRNA | AJ348866.1 | - | 4 | 247 | ||
AK146716.1 | - | 5 | 338 | |||
BC061100.1 | - | 5 | 333 | 30300846 | ||
S70785.1 | - | 4 | 282 |