Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113387.1 | NP_001106858.1 | 3 | 153 | ||
NM_021285.3 | NP_067260.1 | 3 | 330 | |||
GenBank mRNA | AK003182.1 | - | 3 | 306 | ||
AK009938.1 | - | 3 | 142 | |||
BC059087.1 | - | 3 | 120 | 30139109 | ||
BY785713.1 | - | 3 | 155 |