Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031189.2 | NP_112466.1 | 1 | 463 | ||
GenBank mRNA | AB221643.1 | - | 1 | 411 | ||
BC048683.1 | - | 1 | 478 | 6516364 | ||
BC068019.1 | - | 1 | 444 | 6508229 | ||
CJ096370.1 | - | 1 | 463 | |||
D90156.1 | - | 1 | 454 | |||
X15784.1 | - | 1 | 459 |