Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031189.2 | NP_112466.1 | 3 | 640 | ||
GenBank mRNA | AB221643.1 | - | 3 | 588 | ||
BC048683.1 | - | 3 | 655 | 6516364 | ||
BC068019.1 | - | 3 | 621 | 6508229 | ||
D90156.1 | - | 3 | 631 | |||
X15784.1 | - | 3 | 635 |