Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008674.2 | NP_032700.1 | 3 | 302 | ||
GenBank mRNA | AK077364.1 | - | 3 | 302 |