Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010878.2 | NP_035008.2 | 2 | 202 | ||
GenBank mRNA | AF043259.1 | - | 2 | 290 | ||
AF084183.1 | - | 1 | 125 | |||
BC002015.1 | - | 2 | 202 | 3593441 |