Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010894.2 | NP_035024.1 | 2 | 1097 | ||
GenBank mRNA | AK005073.1 | - | 2 | 1098 | ||
AK018781.1 | - | 2 | 1096 | |||
AK028055.1 | - | 2 | 1101 | |||
BC018241.1 | - | 2 | 1073 | 4511370 | ||
BC094611.1 | - | 3 | 679 | 30361598 | ||
U28068.1 | - | 1 | 784 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK044315.1 | - | 51 | Chr. - - [-] on Build |