Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252250.1 | NP_001239179.1 | 8, 9 | 1367 | ||
NM_001252251.1 | NP_001239180.1 | 8, 9 | 1367 | |||
NM_001252252.1 | NP_001239181.1 | 8, 9 | 1367 | |||
NM_001252253.1 | NP_001239182.1 | 8, 9 | 832 | |||
NM_010898.4 | NP_035028.2 | 8, 9 | 1367 | |||
GenBank mRNA | AK017343.1 | - | 8, 9 | 833 | ||
AK045998.1 | - | 8, 9 | 1189 | |||
AK167685.1 | - | 5, 6 | 458 | |||
BC005442.1 | - | 8, 9 | 1233 | 3496053 | ||
DV654856.1 | - | 5, 6 | 425 | |||
L22987.1 | - | 8, 9 | 825 | |||
L22988.1 | - | 8, 9 | 825 | |||
L22989.1 | - | 8, 9 | 825 | |||
L27090.1 | - | 8, 9 | 818 | |||
L27105.1 | - | 8, 9 | 818 | |||
L28176.1 | - | 8, 9 | 1125 | |||
X74671.1 | - | 8, 9 | 1384 |