Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048168.2 | NP_001041633.1 | 4 | 420 | ||
NM_008692.4 | NP_032718.2 | 4 | 493 | |||
GenBank mRNA | AB006181.1 | - | 3 | 273 | ||
AK011327.1 | - | 4 | 450 | |||
BC020117.1 | - | 4 | 419 | 4009737 | ||
BC053723.1 | - | 4 | 423 | 30053020 | ||
BC085261.1 | - | 4 | 419 | 6397760 | ||
BM461330.1 | - | 2 | 124 | |||
CF172077.1 | - | 4 | 416 | |||
CJ092564.1 | - | 4 | 470 | |||
DV650959.1 | - | 4 | 432 |