Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010917.2 | NP_035047.2 | 3 | 773 | ||
GenBank mRNA | AK041633.1 | - | 3 | 709 | ||
AK049585.1 | - | 3 | 709 | |||
AK084876.1 | - | 3 | 712 | |||
AK144878.1 | - | 3 | 738 | |||
AK166779.1 | - | 3 | 709 | |||
BC131669.1 | - | 3 | 684 | 40110859 | ||
X14194.1 | - | 3 | 684 | |||
X14480.1 | - | 3 | 694 |