Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010920.2 | NP_035050.2 | 2 | 718 | ||
GenBank mRNA | AF030113.1 | - | 2 | 531 | ||
AF045150.1 | - | 2 | 718 | |||
AK006401.1 | - | 2 | 574 | |||
AK007304.1 | - | 2 | 501 | |||
BC100425.1 | - | 2 | 536 | |||
BC132558.1 | - | 2 | 568 | 40130835 | ||
BC139035.1 | - | 2 | 568 | 8862057 | ||
BC145516.1 | - | 2 | 572 |