Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010922.2 | NP_035052.2 | 4 | 604 | ||
GenBank mRNA | AF034092.1 | - | 4 | 604 | ||
AK018757.1 | - | 3 | 370 | |||
AK145885.1 | - | 4 | 602 | |||
BC028436.1 | - | 4 | 569 | 1246513 |