Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010924.2 | NP_035054.1 | 2 | 337 | ||
GenBank mRNA | BC028757.1 | - | 2 | 253 | 1067421 | |
CF732455.1 | - | 2 | 337 | |||
U86105.1 | - | 2 | 384 | |||
U86106.1 | - | 2 | 384 | |||
U86107.1 | - | 2 | 384 | |||
U86108.1 | - | 2 | 384 | |||
U89377.1 | - | 2 | 384 |