Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010926.5 | NP_035056.1 | 1, 2 | 355 | ||
GenBank mRNA | AF052621.1 | - | 1, 2 | 357 | ||
AK042704.1 | - | 1, 2 | 223 | |||
AK076710.1 | - | 1, 2 | 818 | |||
AK143131.1 | - | 1, 2 | 316 | |||
AK148190.1 | - | 1, 2 | 255 | |||
AK161438.1 | - | 1, 2 | 331 | |||
AK164130.1 | - | 1, 2 | 356 | |||
BC009103.1 | - | 1, 2 | 311 | 3482942 |