Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010930.4 | NP_035060.1 | 5 | 1040 | ||
GenBank mRNA | AK039481.1 | - | 5 | 1031 | ||
AK081310.1 | - | 5 | 1058 | |||
AK081944.1 | - | 5 | 1266 | |||
AK136407.1 | - | 5 | 1061 | |||
AK155048.1 | - | 5 | 1059 | |||
BC003774.1 | - | 5 | 1046 | 3494896 | ||
X96585.1 | - | 5 | 1040 | |||
Y09257.1 | - | 5 | 1028 |