Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205234.1 | NP_001192163.1 | 2 | 1186 | ||
NM_001205235.1 | NP_001192164.1 | 2 | 1186 | |||
NM_020253.3 | NP_064649.2 | 2 | 1186 | |||
GenBank mRNA | AK129239.1 | - | 2 | 1159 | ||
AK147295.1 | - | 2 | 1121 | |||
AK147665.1 | - | 2 | 1121 | |||
AK162611.1 | - | 2 | 1187 | |||
BC145496.1 | - | 1 | 833 | 9054059 | ||
BC150784.1 | - | 1 | 833 | 9087695 |