Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008747.2 | NP_032773.2 | 2 | 818 | ||
GenBank mRNA | AB041826.1 | - | 2 | 794 | ||
AB041827.1 | - | 2 | 794 | |||
AK005368.1 | - | 2 | 533 | |||
AK081073.1 | - | 2 | 551 | |||
AK158943.1 | - | 2 | 818 | |||
BC141019.1 | - | 2 | 813 | 9055631 | ||
U51908.1 | - | 2 | 817 |
Set Membership: |
G-protein coupled receptor |