Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010950.2 | NP_035080.2 | 2 | 219 | ||
GenBank mRNA | AK147523.1 | - | 1 | 3383 | ||
BC059915.1 | - | 1 | 808 | |||
BC068116.1 | - | 2 | 46 | 6809443 | ||
BC098097.1 | - | 2 | 60 | 6403012 | ||
CJ070714.1 | - | 2 | 219 | |||
U96441.1 | - | 2 | 103 |