Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sebox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008759.2 | NP_032785.1 | 3 | 392 | ||
GenBank mRNA | AK139838.1 | - | 3 | 416 | ||
AK143429.1 | - | 3 | 404 | |||
BC098184.1 | - | 3 | 392 | 5367903 |