Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146934.2 | NP_667145.1 | 3 | 675 | ||
GenBank mRNA | AF102520.1 | - | 1 | 229 | ||
AF102526.1 | - | 1 | 229 | |||
AF102531.1 | - | 229 | ||||
BC108947.1 | - | 1 | 440 | 40043808 |