Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011026.2 | NP_035156.2 | 3 | 434 | ||
GenBank mRNA | AF089751.1 | - | 3 | 368 | ||
AF089752.1 | - | 3 | 368 | |||
AJ251459.1 | - | 3 | 348 | |||
AJ251460.1 | - | 3 | 348 | |||
AJ251461.1 | - | 3 | 348 | |||
AJ251462.1 | - | 3 | 348 | |||
AK150351.1 | - | 3 | 424 | |||
AK162820.1 | - | 3 | 400 | |||
AK163104.1 | - | 3 | 422 | |||
BC002099.1 | - | 3 | 407 | |||
BC005597.1 | - | 3 | 382 | 3482458 | ||
JX417451.1 | - | 3 | 394 |