Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038839.2 | NP_001033928.1 | 1 | 227 | ||
NM_001038845.2 | NP_001033934.1 | 1 | 227 | |||
NM_001038887.1 | NP_001033976.1 | 1 | 227 | |||
NM_011027.3 | NP_035157.2 | 1 | 227 | |||
GenBank mRNA | AJ009823.1 | - | 1 | 75 | ||
AJ489296.1 | - | 1 | 64 | |||
AJ489297.1 | - | 1 | 64 | |||
AK144585.1 | - | 1 | 228 | |||
AK145050.1 | - | 1 | 82 | |||
AK145211.1 | - | 1 | 91 | |||
AK171132.1 | - | 1 | 199 | |||
BC062808.1 | - | 1 | 129 | |||
BC128303.1 | - | 1 | 71 | |||
BC128304.1 | - | 1 | 71 | |||
BC141120.1 | - | 1 | 94 | 9055732 | ||
BY752747.1 | - | 1 | 169 |