Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013625.4 | NP_038653.1 | 3 | 640 | ||
GenBank mRNA | AK144908.1 | - | 3 | 629 | ||
AK147207.1 | - | 3 | 578 | |||
AK147229.1 | - | 3 | 591 | |||
AK147237.1 | - | 3 | 632 | |||
AK147309.1 | - | 3 | 575 | |||
AK147507.1 | - | 3 | 914 | |||
AK147515.1 | - | 3 | 627 | |||
AK147533.1 | - | 3 | 607 | |||
AK147793.1 | - | 3 | 302 | |||
AK160764.1 | - | 3 | 857 | |||
AK164008.1 | - | 3 | 311 | |||
AK167448.1 | - | 3 | 607 | |||
AK167824.1 | - | 3 | 632 | |||
AK168929.1 | - | 3 | 280 | |||
AK168960.1 | - | 3 | 595 | |||
AY189217.1 | - | 2 | 259 | |||
AY189218.1 | - | 2 | 259 | |||
BC014831.1 | - | 3 | 579 | 4507474 | ||
BC026141.1 | - | 3 | 604 | 4017963 | ||
BC046989.1 | - | 3 | 613 | |||
BC099462.1 | - | 3 | 579 | |||
BQ895724.1 | - | 3 | 640 | |||
L25108.1 | - | 3 | 540 | |||
U95116.1 | - | 3 | 935 |