Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013625.4 | NP_038653.1 | 3, 4 | 670 | ||
GenBank mRNA | AK144908.1 | - | 3, 4 | 659 | ||
AK147207.1 | - | 3, 4 | 608 | |||
AK147229.1 | - | 3, 4 | 621 | |||
AK147237.1 | - | 3, 4 | 662 | |||
AK147309.1 | - | 3, 4 | 605 | |||
AK147507.1 | - | 3, 4 | 944 | |||
AK147515.1 | - | 3, 4 | 657 | |||
AK147533.1 | - | 3, 4 | 637 | |||
AK147793.1 | - | 3, 4 | 332 | |||
AK160764.1 | - | 3, 4 | 887 | |||
AK164008.1 | - | 3, 4 | 341 | |||
AK167448.1 | - | 3, 4 | 637 | |||
AK167824.1 | - | 3, 4 | 662 | |||
AK168929.1 | - | 3, 4 | 310 | |||
AK168960.1 | - | 3, 4 | 625 | |||
AY189217.1 | - | 2, 3 | 289 | |||
AY189218.1 | - | 2, 3 | 289 | |||
BC014831.1 | - | 3, 4 | 609 | 4507474 | ||
BC026141.1 | - | 3, 4 | 634 | 4017963 | ||
BC046989.1 | - | 3, 4 | 643 | |||
BC099462.1 | - | 3, 4 | 609 | |||
BQ895724.1 | - | 3, 4 | 670 | |||
L25108.1 | - | 3, 4 | 570 | |||
U95116.1 | - | 3, 4 | 965 |